Product Details

SNP ID
rs149718842
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:43061414 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAGAGGAGATCCTGGGGAGCACA[C/T]GGCTGGTCAGCCAAGGGCTAGAGGC
Phenotype
MIM: 609577 MIM: 611822
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CUL7 PubMed Links

Gene Details

Gene
CUL7
Gene Name
cullin 7
There are no transcripts associated with this gene.

Gene
KLC4
Gene Name
kinesin light chain 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289034.1 196 Missense Mutation CGG,TGG R27W NP_001275963.1
NM_001289035.1 196 Missense Mutation CGG,TGG R27W NP_001275964.1
NM_138343.3 196 Missense Mutation CGG,TGG R27W NP_612352.1
NM_201521.2 196 Missense Mutation CGG,TGG R27W NP_958929.1
NM_201522.2 196 Missense Mutation CGG,TGG R27W NP_958930.1
NM_201523.2 196 Missense Mutation CGG,TGG R45W NP_958931.1
XM_011514989.1 196 Missense Mutation CGG,TGG R45W XP_011513291.1
XM_017011457.1 196 Missense Mutation CGG,TGG R27W XP_016866946.1
Gene
MRPL2
Gene Name
mitochondrial ribosomal protein L2
There are no transcripts associated with this gene.

View Full Product Details