Product Details

SNP ID
rs149728484
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:109979430 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCCACTCCGGCGCTGCGCACGCC[C/T]ATGTTCGTGCTGGTAGGCAGCCTGG
Phenotype
MIM: 600553
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR6 PubMed Links

Gene Details

Gene
GPR6
Gene Name
G protein-coupled receptor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286099.1 657 Silent Mutation CCC,CCT P121P NP_001273028.1
NM_005284.4 657 Silent Mutation CCC,CCT P106P NP_005275.1

View Full Product Details