Product Details
- SNP ID
-
rs149937283
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:26107560 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCTTGCCTTCACTGGGCTCTTCTG[C/T]TGTTGCTTACCCTTGGCTCCTTTAG
- Phenotype
-
MIM: 142712
MIM: 602827
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HIST1H1T
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs198845] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HIST1H1T
- Gene Name
- histone cluster 1, H1t
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005323.3 |
577 |
Silent Mutation |
|
|
NP_005314.2 |
- Gene
- HIST1H4C
- Gene Name
- histone cluster 1, H4c
There are no transcripts associated with this gene.
View Full Product Details