Product Details

SNP ID
rs149942995
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:27249132 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCAGCCTTGGCCCCAGCCTGGGGC[A/G]CCCTGGTGATAAGCCTGGAACACAG
Phenotype
MIM: 607169
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRSS16 PubMed Links

Gene Details

Gene
PRSS16
Gene Name
protease, serine 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005865.3 385 Missense Mutation ACC,GCC T124A NP_005856.1
XM_017010161.1 385 Missense Mutation ACC,GCC T124A XP_016865650.1
XM_017010162.1 385 Missense Mutation ACC,GCC T124A XP_016865651.1
XM_017010163.1 385 Missense Mutation ACC,GCC T124A XP_016865652.1
XM_017010164.1 385 Missense Mutation ACC,GCC T124A XP_016865653.1
XM_017010165.1 385 Intron XP_016865654.1

View Full Product Details