Product Details

SNP ID
rs150035394
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:75088078 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACCTGAAGGAAACAGCTCTGTTA[A/C]GCAGCTACATTGAGCTGCCATCAAA
Phenotype
MIM: 120320
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
COL12A1 PubMed Links

Gene Details

Gene
COL12A1
Gene Name
collagen type XII alpha 1 chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004370.5 Intron NP_004361.3
NM_080645.2 Intron NP_542376.2
XM_011535434.1 Intron XP_011533736.1
XM_011535435.1 Intron XP_011533737.1
XM_011535436.2 Intron XP_011533738.1
XM_017010252.1 Intron XP_016865741.1

View Full Product Details