Product Details

SNP ID
rs150277350
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:41158975 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATGCCAGGCTGCAGCCCAGAGGG[C/T]GCTGGCTGCTAGAATCTTGATGAGA
Phenotype
MIM: 605086 MIM: 609714
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
LOC105375056 PubMed Links

Gene Details

Gene
LOC105375056
Gene Name
uncharacterized LOC105375056
There are no transcripts associated with this gene.

Gene
TREM2
Gene Name
triggering receptor expressed on myeloid cells 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271821.1 678 Intron NP_001258750.1
NM_018965.3 678 Missense Mutation ACC,GCC T192A NP_061838.1
XM_006715116.3 678 Missense Mutation ACC,GCC T105A XP_006715179.1
Gene
TREML1
Gene Name
triggering receptor expressed on myeloid cells like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271807.1 678 Intron NP_001258736.1
NM_001271808.1 678 Intron NP_001258737.1
NM_178174.3 678 Intron NP_835468.1
XM_017010822.1 678 Intron XP_016866311.1
XM_017010823.1 678 Intron XP_016866312.1
XM_017010824.1 678 Intron XP_016866313.1
XM_017010825.1 678 Intron XP_016866314.1

View Full Product Details