Product Details

SNP ID
rs150333384
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:73431532 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGATGAAAGCAGAACATGCTAAA[C/T]GACACTTCTAGAATGCACTCTACAA
Phenotype
MIM: 613973
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MB21D1 PubMed Links

Gene Details

Gene
MB21D1
Gene Name
Mab-21 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138441.2 Intron NP_612450.2
XM_017010232.1 Intron XP_016865721.1

View Full Product Details