Product Details
- SNP ID
-
rs150356042
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:132617677 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCATAGGCCTCTGAGAAGACCACCC[C/T]GAAGGCAAATGCTCCAGGGACCGAC
- Phenotype
-
MIM: 604849
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TAAR2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs61745666] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TAAR2
- Gene Name
- trace amine associated receptor 2
- Gene
- TAAR3
- Gene Name
- trace amine associated receptor 3 (gene/pseudogene)
There are no transcripts associated with this gene.
View Full Product Details