Product Details

SNP ID
rs150416507
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:28124643 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTGTTCGGGGCTGTCCAAAGGAC[G/T]CTAGCTGTTGCACCTGTTCCTCCCT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZSCAN16 PubMed Links

Gene Details

Gene
ZSCAN16
Gene Name
zinc finger and SCAN domain containing 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320555.1 88 UTR 5 NP_001307484.1
NM_001320556.1 88 UTR 5 NP_001307485.1
NM_001320557.1 88 UTR 5 NP_001307486.1
NM_001320558.1 88 UTR 5 NP_001307487.1
NM_025231.2 88 UTR 5 NP_079507.1
XM_017011324.1 88 UTR 5 XP_016866813.1
Gene
ZSCAN16-AS1
Gene Name
ZSCAN16 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details