Product Details

SNP ID
rs150463278
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30601576 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGCAGGGGGGGCCCATTGACACC[C/T]GGGTGGTAGAAGGCACAGTTGTTCT
Phenotype
MIM: 603771
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
PPP1R10 PubMed Links

Gene Details

Gene
PPP1R10
Gene Name
protein phosphatase 1 regulatory subunit 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002714.3 3402 Silent Mutation CCA,CCG P932P NP_002705.2
XM_006715130.2 3402 Silent Mutation CCA,CCG P932P XP_006715193.1
XM_011514722.1 3402 Silent Mutation CCA,CCG P932P XP_011513024.1
XM_017010995.1 3402 Silent Mutation CCA,CCG P932P XP_016866484.1

View Full Product Details