Product Details

SNP ID
rs150485975
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:119179839 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTTTATTCCTCTCTGATTTCAA[C/G]TTCCTTTTTATCTTTAGGGAGGATA
Phenotype
MIM: 604344
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MAN1A1 PubMed Links

Gene Details

Gene
MAN1A1
Gene Name
mannosidase alpha class 1A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005907.3 2245 Missense Mutation CTT,GTT L648V NP_005898.2
XM_005266986.4 2245 Missense Mutation CTT,GTT L731V XP_005267043.1
XM_011535833.2 2245 Missense Mutation CTT,GTT L459V XP_011534135.1
XM_011535834.2 2245 Intron XP_011534136.1

View Full Product Details