Product Details
- SNP ID
-
rs150560383
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:25284831 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTCAGAAAGCATAAAGGATGTTAT[C/T]GGCAGAAAGATAAAAATTTCAGTGA
- Phenotype
-
MIM: 609593
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CARMIL1
PubMed Links
Gene Details
- Gene
- CARMIL1
- Gene Name
- capping protein regulator and myosin 1 linker 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001173977.1 |
428 |
Silent Mutation |
ATC,ATT |
I20I |
NP_001167448.1 |
NM_017640.5 |
428 |
Silent Mutation |
ATC,ATT |
I20I |
NP_060110.4 |
XM_005249218.1 |
428 |
Silent Mutation |
ATC,ATT |
I20I |
XP_005249275.1 |
XM_017011008.1 |
428 |
Silent Mutation |
ATC,ATT |
I20I |
XP_016866497.1 |
XM_017011009.1 |
428 |
Silent Mutation |
ATC,ATT |
I20I |
XP_016866498.1 |
XM_017011010.1 |
428 |
Silent Mutation |
ATC,ATT |
I20I |
XP_016866499.1 |
XM_017011011.1 |
428 |
Silent Mutation |
ATC,ATT |
I20I |
XP_016866500.1 |
XM_017011012.1 |
428 |
Silent Mutation |
ATC,ATT |
I20I |
XP_016866501.1 |
XM_017011013.1 |
428 |
Silent Mutation |
ATC,ATT |
I20I |
XP_016866502.1 |
XM_017011014.1 |
428 |
Silent Mutation |
ATC,ATT |
I20I |
XP_016866503.1 |
XM_017011015.1 |
428 |
Silent Mutation |
ATC,ATT |
I20I |
XP_016866504.1 |
XM_017011016.1 |
428 |
Silent Mutation |
ATC,ATT |
I20I |
XP_016866505.1 |
XM_017011017.1 |
428 |
Silent Mutation |
ATC,ATT |
I20I |
XP_016866506.1 |
- Gene
- LOC105375106
- Gene Name
- uncharacterized LOC105375106
There are no transcripts associated with this gene.
View Full Product Details