Product Details

SNP ID
rs150775105
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:75926877 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGGAAAAATAAAGGTGAAAATGT[A/G]ATGAGAAACAATGACAAGTAACTGA
Phenotype
MIM: 602870 MIM: 600970
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IMPG1 PubMed Links

Gene Details

Gene
IMPG1
Gene Name
interphotoreceptor matrix proteoglycan 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282368.1 Intron NP_001269297.1
NM_001563.3 Intron NP_001554.2
Gene
MYO6
Gene Name
myosin VI
There are no transcripts associated with this gene.

View Full Product Details