Product Details

SNP ID
rs151009303
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:52978324 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAAAGGTTGATATTTCTAGAAGA[A/G]ATGATCTCGTTGACACAAAAGACCC
Phenotype
MIM: 605450
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GSTA4 PubMed Links

Gene Details

Gene
GSTA4
Gene Name
glutathione S-transferase alpha 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001512.3 965 UTR 3 NP_001503.1
XM_005249035.3 965 UTR 3 XP_005249092.1
XM_011514534.2 965 UTR 3 XP_011512836.1
XM_011514535.2 965 UTR 3 XP_011512837.1

View Full Product Details