Product Details

SNP ID
rs151192215
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31672206 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTTGTCTTTTGCTGAGCTGCGCC[A/G]CATGTACTTGTTCCTCAATAGTTCA
Phenotype
MIM: 115441 MIM: 142610 MIM: 610433 MIM: 610434
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CSNK2B PubMed Links

Gene Details

Gene
CSNK2B
Gene Name
casein kinase 2 beta
There are no transcripts associated with this gene.

Gene
GPANK1
Gene Name
G-patch domain and ankyrin repeats 1
There are no transcripts associated with this gene.

Gene
LY6G5B
Gene Name
lymphocyte antigen 6 complex, locus G5B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021221.2 530 Missense Mutation CAC,CGC H177R NP_067044.2
Gene
LY6G5C
Gene Name
lymphocyte antigen 6 complex, locus G5C
There are no transcripts associated with this gene.

View Full Product Details