Product Details

SNP ID
rs151253133
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:41739826 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCATACTCATCCTCCTGGGCCCC[C/T]GTGGCCTGCAGAAGAGCACTCATGT
Phenotype
MIM: 169740 MIM: 600744
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PGC PubMed Links

Gene Details

Gene
PGC
Gene Name
progastricsin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166424.1 955 Intron NP_001159896.1
NM_002630.3 955 Silent Mutation ACA,ACG T296T NP_002621.1
Gene
TFEB
Gene Name
transcription factor EB
There are no transcripts associated with this gene.

View Full Product Details