Product Details

SNP ID
rs151258622
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:152972406 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGAAGCCAGTGGGGTTCTGAACA[A/T]AACATATTCTCTGGTTGAAGCATGA
Phenotype
MIM: 606013
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FBXO5 PubMed Links

Gene Details

Gene
FBXO5
Gene Name
F-box protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142522.1 1067 Missense Mutation ATG,TTG M274L NP_001135994.1
NM_012177.3 1067 Missense Mutation ATG,TTG M320L NP_036309.1

View Full Product Details