Product Details

SNP ID
rs137864032
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:28953726 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAATCTAAAATAAGCCCTGGCAG[A/C]AAACTAAGAATGACAGTCTATTTCC
Phenotype
MIM: 613356
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TRIL PubMed Links

Gene Details

Gene
TRIL
Gene Name
TLR4 interactor with leucine rich repeats
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014817.3 4688 UTR 3 NP_055632.2

View Full Product Details