Product Details
- SNP ID
-
rs138167675
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:151557207 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCGTCTACATTCACGGCGGTCACTC[C/T]GTTTCTGTCTCCTTTTGTTTGGCAC
- Phenotype
-
MIM: 602743
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PRKAG2
PubMed Links
Gene Details
- Gene
- PRKAG2
- Gene Name
- protein kinase AMP-activated non-catalytic subunit gamma 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001040633.1 |
2080 |
Silent Mutation |
ACA,ACG |
T524T |
NP_001035723.1 |
NM_001304527.1 |
2080 |
Silent Mutation |
ACA,ACG |
T443T |
NP_001291456.1 |
NM_001304531.1 |
2080 |
Silent Mutation |
ACA,ACG |
T327T |
NP_001291460.1 |
NM_016203.3 |
2080 |
Silent Mutation |
ACA,ACG |
T568T |
NP_057287.2 |
NM_024429.1 |
2080 |
Silent Mutation |
ACA,ACG |
T327T |
NP_077747.1 |
XM_005250002.3 |
2080 |
UTR 3 |
|
|
XP_005250059.1 |
XM_005250004.3 |
2080 |
UTR 3 |
|
|
XP_005250061.1 |
XM_005250006.4 |
2080 |
UTR 3 |
|
|
XP_005250063.1 |
XM_006716021.2 |
2080 |
UTR 3 |
|
|
XP_006716084.1 |
XM_011516282.1 |
2080 |
UTR 3 |
|
|
XP_011514584.1 |
XM_011516283.1 |
2080 |
Silent Mutation |
ACA,ACG |
T564T |
XP_011514585.1 |
XM_011516284.1 |
2080 |
Silent Mutation |
ACA,ACG |
T563T |
XP_011514586.1 |
XM_011516285.1 |
2080 |
UTR 3 |
|
|
XP_011514587.1 |
XM_011516286.1 |
2080 |
UTR 3 |
|
|
XP_011514588.1 |
XM_011516287.1 |
2080 |
UTR 3 |
|
|
XP_011514589.1 |
XM_017012268.1 |
2080 |
UTR 3 |
|
|
XP_016867757.1 |
XM_017012269.1 |
2080 |
Silent Mutation |
ACA,ACG |
T567T |
XP_016867758.1 |
XM_017012270.1 |
2080 |
Silent Mutation |
ACA,ACG |
T524T |
XP_016867759.1 |
XM_017012271.1 |
2080 |
Silent Mutation |
ACA,ACG |
T523T |
XP_016867760.1 |
XM_017012272.1 |
2080 |
Silent Mutation |
ACA,ACG |
T523T |
XP_016867761.1 |
XM_017012273.1 |
2080 |
Silent Mutation |
ACA,ACG |
T444T |
XP_016867762.1 |
XM_017012274.1 |
2080 |
UTR 3 |
|
|
XP_016867763.1 |
XM_017012275.1 |
2080 |
UTR 3 |
|
|
XP_016867764.1 |
XM_017012276.1 |
2080 |
Silent Mutation |
ACA,ACG |
T326T |
XP_016867765.1 |
XM_017012277.1 |
2080 |
Silent Mutation |
ACA,ACG |
T319T |
XP_016867766.1 |
XM_017012278.1 |
2080 |
Silent Mutation |
ACA,ACG |
T307T |
XP_016867767.1 |
XM_017012279.1 |
2080 |
Silent Mutation |
ACA,ACG |
T307T |
XP_016867768.1 |
XM_017012280.1 |
2080 |
Silent Mutation |
ACA,ACG |
T307T |
XP_016867769.1 |
XM_017012281.1 |
2080 |
Silent Mutation |
ACA,ACG |
T307T |
XP_016867770.1 |
XM_017012282.1 |
2080 |
Silent Mutation |
ACA,ACG |
T307T |
XP_016867771.1 |
View Full Product Details