Product Details

SNP ID
rs138324715
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:102457730 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCGGGCGCCTCCCGACACATGGA[C/T]AGCACGGTGGGGGACAGGAGGTTGA
Phenotype
MIM: 613302 MIM: 615167 MIM: 610929
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALKBH4 PubMed Links

Gene Details

Gene
ALKBH4
Gene Name
alkB homolog 4, lysine demethylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017621.3 611 Silent Mutation CTA,CTG L191L NP_060091.1
XM_005250464.2 611 Silent Mutation CTA,CTG L118L XP_005250521.1
XM_005250465.2 611 Silent Mutation CTA,CTG L53L XP_005250522.1
Gene
LRWD1
Gene Name
leucine rich repeats and WD repeat domain containing 1
There are no transcripts associated with this gene.

Gene
MIR5090
Gene Name
microRNA 5090
There are no transcripts associated with this gene.

Gene
ORAI2
Gene Name
ORAI calcium release-activated calcium modulator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126340.2 611 Intron NP_001119812.1
NM_001271818.1 611 Intron NP_001258747.1
NM_001271819.1 611 Intron NP_001258748.1
NM_032831.3 611 Intron NP_116220.1

View Full Product Details