Product Details

SNP ID
rs138329092
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:77196958 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTTGCCTATTGCGTAGTAGTCA[C/G]AGCAATCTTTATATATTAGATGTTG
Phenotype
MIM: 605351
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCDC146 PubMed Links

Gene Details

Gene
CCDC146
Gene Name
coiled-coil domain containing 146
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020879.2 681 Intron NP_065930.2
Gene
FGL2
Gene Name
fibrinogen like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006682.2 681 Missense Mutation TCT,TGT S214C NP_006673.1

View Full Product Details