Product Details

SNP ID
rs138458340
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:112821553 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGCTATGACATGAGTCACTCTGC[A/G]TGAAACTTTAAGCTTTCTATATAAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BMT2 PubMed Links

Gene Details

Gene
BMT2
Gene Name
base methyltransferase of 25S rRNA 2 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152556.2 1536 UTR 3 NP_689769.2

View Full Product Details