Product Details

SNP ID
rs138497833
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:76400311 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCACACAGGACAGCCACATCCTC[A/G]TAGTGAAAGCAATTGTGGACGCCCC
Phenotype
MIM: 607639 MIM: 182889
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SSC4D PubMed Links

Gene Details

Gene
SSC4D
Gene Name
scavenger receptor cysteine rich family member with 4 domains
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080744.1 798 Silent Mutation TAC,TAT Y150Y NP_542782.1
XM_017011750.1 798 Intron XP_016867239.1
Gene
ZP3
Gene Name
zona pellucida glycoprotein 3 (sperm receptor)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001110354.1 798 Intron NP_001103824.1
NM_007155.5 798 Intron NP_009086.4
XM_017012586.1 798 Intron XP_016868075.1

View Full Product Details