Product Details

SNP ID
rs138569406
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:148810257 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCACCTCACTGACCTCTACCCTC[A/G]TTTCTGAACACTCGGCCGGCAGAAG
Phenotype
MIM: 603134 MIM: 601573
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CUL1 PubMed Links

Gene Details

Gene
CUL1
Gene Name
cullin 1
There are no transcripts associated with this gene.

Gene
EZH2
Gene Name
enhancer of zeste 2 polycomb repressive complex 2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001203247.1 Intron NP_001190176.1
NM_001203248.1 Intron NP_001190177.1
NM_001203249.1 Intron NP_001190178.1
NM_004456.4 Intron NP_004447.2
NM_152998.2 Intron NP_694543.1
XM_005249962.4 Intron XP_005250019.1
XM_005249963.4 Intron XP_005250020.1
XM_005249964.4 Intron XP_005250021.1
XM_011515883.2 Intron XP_011514185.1
XM_011515884.2 Intron XP_011514186.1
XM_011515885.2 Intron XP_011514187.1
XM_011515886.2 Intron XP_011514188.1
XM_011515887.2 Intron XP_011514189.1
XM_011515888.2 Intron XP_011514190.1
XM_011515889.2 Intron XP_011514191.1
XM_011515890.2 Intron XP_011514192.1
XM_011515891.2 Intron XP_011514193.1
XM_011515892.2 Intron XP_011514194.1
XM_011515893.2 Intron XP_011514195.1
XM_011515894.2 Intron XP_011514196.1
XM_011515895.2 Intron XP_011514197.1
XM_011515896.2 Intron XP_011514198.1
XM_011515897.2 Intron XP_011514199.1
XM_011515898.2 Intron XP_011514200.1
XM_011515899.2 Intron XP_011514201.1
XM_011515901.2 Intron XP_011514203.1
XM_017011817.1 Intron XP_016867306.1
XM_017011818.1 Intron XP_016867307.1
XM_017011819.1 Intron XP_016867308.1
XM_017011820.1 Intron XP_016867309.1
XM_017011821.1 Intron XP_016867310.1

View Full Product Details