Product Details

SNP ID
rs138579798
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:149198353 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACAGGCTCAAGAATGGGACATGGA[C/T]GCCCGGCGGCCAATGCCTTTTCAGT
Phenotype
MIM: 603397
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF282 PubMed Links

Gene Details

Gene
ZNF282
Gene Name
zinc finger protein 282
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303481.1 314 Silent Mutation GAC,GAT D62D NP_001290410.1
NM_003575.3 314 Silent Mutation GAC,GAT D62D NP_003566.1
XM_006716151.3 314 Silent Mutation GAC,GAT D62D XP_006716214.1

View Full Product Details