Product Details

SNP ID
rs138768813
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:151192703 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTTCCAGCTTAGCCCCAGCTCAC[A/G]CTACCTGCCCACACGGCCTTCACAT
Phenotype
MIM: 615054
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASB10 PubMed Links

Gene Details

Gene
ASB10
Gene Name
ankyrin repeat and SOCS box containing 10
There are no transcripts associated with this gene.

Gene
IQCA1L
Gene Name
IQ motif containing with AAA domain 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304419.1 Intron NP_001291348.1

View Full Product Details