Product Details

SNP ID
rs138791795
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:93926134 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAATAAAGAACTATATTGAAGAAC[A/G]TTCTGGAGAGGATCCTCTAGTAAAG
Phenotype
MIM: 604390
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GNG11 PubMed Links

Gene Details

Gene
GNG11
Gene Name
G protein subunit gamma 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004126.3 574 Missense Mutation CAT,CGT H47R NP_004117.1

View Full Product Details