Product Details

SNP ID
rs138819536
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:41690245 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGCTCTTGCCCTGGTCCAGCAAC[C/T]GCTGGATGCTGCTGGAGACAGGGAA
Phenotype
MIM: 147290
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
INHBA PubMed Links

Gene Details

Gene
INHBA
Gene Name
inhibin beta A subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017012174.1 2693 Missense Mutation XP_016867663.1
XM_017012175.1 2693 Missense Mutation XP_016867664.1
XM_017012176.1 2693 Missense Mutation XP_016867665.1
Gene
INHBA-AS1
Gene Name
INHBA antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details