Product Details

SNP ID
rs138945257
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:21901049 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGCAGGAACCATTGTTGAAGCCC[A/G]TCTCAAGGAGCTGGCATGCCCTATG
Phenotype
MIM: 609685 MIM: 603339
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CDCA7L PubMed Links

Gene Details

Gene
CDCA7L
Gene Name
cell division cycle associated 7 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127370.2 2758 UTR 3 NP_001120842.1
NM_001127371.2 2758 UTR 3 NP_001120843.1
NM_018719.4 2758 UTR 3 NP_061189.2
Gene
DNAH11
Gene Name
dynein axonemal heavy chain 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277115.1 2758 Missense Mutation CAT,CGT H4449R NP_001264044.1

View Full Product Details