Product Details

SNP ID
rs139200164
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:151155042 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTCCCTCATTCACCCCATTACTG[C/T]AGCTCCTGCTGTTATCACCGTAGGA
Phenotype
MIM: 603354
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GBX1 PubMed Links

Gene Details

Gene
GBX1
Gene Name
gastrulation brain homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098834.2 Intron NP_001092304.1
XM_017011964.1 Intron XP_016867453.1
XM_017011965.1 Intron XP_016867454.1

View Full Product Details