Product Details

SNP ID
rs139212136
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:101222767 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTGCCTCAGTTTGATGACGATGC[A/G]GACACTGGTGAGGCGGATGGAGGAG
Phenotype
MIM: 615778 MIM: 603066
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLDN15 PubMed Links

Gene Details

Gene
CLDN15
Gene Name
claudin 15
There are no transcripts associated with this gene.

Gene
PLOD3
Gene Name
procollagen-lysine,2-oxoglutarate 5-dioxygenase 3
There are no transcripts associated with this gene.

Gene
ZNHIT1
Gene Name
zinc finger HIT-type containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006349.2 678 Silent Mutation GCA,GCG A62A NP_006340.1
XM_011515739.2 678 Silent Mutation GCA,GCG A77A XP_011514041.1

View Full Product Details