Product Details
- SNP ID
-
rs139406164
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:140335468 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCACGAGAGAGAATATTTCCCTCA[C/G]TATTAATGGCGAGATAAACACAATT
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SLC37A3
PubMed Links
Gene Details
- Gene
- SLC37A3
- Gene Name
- solute carrier family 37 member 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001287498.1 |
1930 |
Missense Mutation |
CTG,GTG |
L461V |
NP_001274427.1 |
NM_032295.3 |
1930 |
Missense Mutation |
ACT,AGT |
T376S |
NP_115671.1 |
NM_207113.2 |
1930 |
Missense Mutation |
CTG,GTG |
L477V |
NP_996996.1 |
XM_011516626.2 |
1930 |
Missense Mutation |
ACT,AGT |
T426S |
XP_011514928.1 |
XM_011516627.2 |
1930 |
Missense Mutation |
ACT,AGT |
T410S |
XP_011514929.1 |
XM_017012712.1 |
1930 |
Missense Mutation |
CTG,GTG |
L477V |
XP_016868201.1 |
XM_017012713.1 |
1930 |
Missense Mutation |
ACT,AGT |
T376S |
XP_016868202.1 |
XM_017012714.1 |
1930 |
Missense Mutation |
CTG,GTG |
L396V |
XP_016868203.1 |
XM_017012715.1 |
1930 |
Missense Mutation |
CTG,GTG |
L389V |
XP_016868204.1 |
XM_017012716.1 |
1930 |
Missense Mutation |
ACT,AGT |
T288S |
XP_016868205.1 |
XM_017012717.1 |
1930 |
Missense Mutation |
CTG,GTG |
L329V |
XP_016868206.1 |
XM_017012718.1 |
1930 |
Intron |
|
|
XP_016868207.1 |
View Full Product Details