Product Details

SNP ID
rs139412585
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:128028791 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCCCCAGGCTGTTTTCTGTCCAG[C/T]GGGCCCCATGTGCTGGTTTGTAGGT
Phenotype
MIM: 610486 MIM: 602181
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRRC4 PubMed Links

Gene Details

Gene
LRRC4
Gene Name
leucine rich repeat containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022143.4 2293 Missense Mutation CAC,CGC H617R NP_071426.1
XM_011516461.2 2293 Missense Mutation CAC,CGC H617R XP_011514763.1
XM_017012505.1 2293 Missense Mutation CAC,CGC H617R XP_016867994.1
Gene
SND1
Gene Name
staphylococcal nuclease and tudor domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017011987.1 2293 Intron XP_016867476.1

View Full Product Details