Product Details

SNP ID
rs139581191
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:27147325 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCTTACCCGCGCAGGAGTTCATC[C/T]GCTGCATCCAAGGGTAAACCGGGCT
Phenotype
MIM: 142954 MIM: 142952 MIM: 142951 MIM: 142950
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HOXA-AS3 PubMed Links

Gene Details

Gene
HOXA-AS3
Gene Name
HOXA cluster antisense RNA 3
There are no transcripts associated with this gene.

Gene
HOXA3
Gene Name
homeobox A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030661.4 450 Intron NP_109377.1
NM_153631.2 450 Intron NP_705895.1
XM_005249730.2 450 Intron XP_005249787.1
XM_005249731.2 450 Intron XP_005249788.1
XM_005249732.3 450 Intron XP_005249789.1
XM_006715715.2 450 Intron XP_006715778.1
XM_011515343.2 450 Intron XP_011513645.1
Gene
HOXA5
Gene Name
homeobox A5
There are no transcripts associated with this gene.

Gene
HOXA6
Gene Name
homeobox A6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024014.3 450 Missense Mutation CAG,CGG Q142R NP_076919.1
Gene
HOXA7
Gene Name
homeobox A7
There are no transcripts associated with this gene.

View Full Product Details