Product Details
- SNP ID
-
rs139681941
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:91265951 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTACCCGGAGCGGCCCATCATCTT[C/T]TTGTCCGGCTGTTACACGGCCGTGG
- Phenotype
-
MIM: 603408
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FZD1
PubMed Links
Gene Details
- Gene
- FZD1
- Gene Name
- frizzled class receptor 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003505.1 |
1484 |
Silent Mutation |
TTC,TTT |
F357F |
NP_003496.1 |
View Full Product Details