Product Details

SNP ID
rs139782938
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:41963387 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAGATGGAACTAATCAACAAACA[C/T]AGCAACTCATAAATATTCCCCCAAC
Phenotype
MIM: 165240
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GLI3 PubMed Links

Gene Details

Gene
GLI3
Gene Name
GLI family zinc finger 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000168.5 5777 UTR 3 NP_000159.3
XM_011515274.2 5777 Intron XP_011513576.1
XM_017011997.1 5777 Intron XP_016867486.1

View Full Product Details