Product Details
- SNP ID
-
rs139785079
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:108479293 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATAACGTCCAGTGCCCAGGGATAC[C/T]ATGCACTCTAACGGCACATCTGGCC
- Phenotype
-
MIM: 612123
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PNPLA8
PubMed Links
Gene Details
- Gene
- PNPLA8
- Gene Name
- patatin like phospholipase domain containing 8
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001256007.2 |
3213 |
Missense Mutation |
ATA,ATG |
I655M |
NP_001242936.1 |
NM_001256008.2 |
3213 |
Missense Mutation |
ATA,ATG |
I655M |
NP_001242937.1 |
NM_001256009.2 |
3213 |
Missense Mutation |
ATA,ATG |
I593M |
NP_001242938.1 |
NM_001256010.2 |
3213 |
Missense Mutation |
ATA,ATG |
I555M |
NP_001242939.1 |
NM_001256011.2 |
3213 |
Missense Mutation |
ATA,ATG |
I555M |
NP_001242940.1 |
NM_015723.4 |
3213 |
Missense Mutation |
ATA,ATG |
I655M |
NP_056538.1 |
XM_005250396.4 |
3213 |
Missense Mutation |
ATA,ATG |
I593M |
XP_005250453.1 |
XM_011516274.2 |
3213 |
Missense Mutation |
ATA,ATG |
I683M |
XP_011514576.1 |
XM_011516275.2 |
3213 |
Missense Mutation |
ATA,ATG |
I621M |
XP_011514577.1 |
XM_017012261.1 |
3213 |
Missense Mutation |
ATA,ATG |
I493M |
XP_016867750.1 |
View Full Product Details