Product Details

SNP ID
rs139973261
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:141972720 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGTCCGGGAATCTGCCTTGTGGT[C/T]GGCTCTTACCTTCAGGCTGCTCTGA
Phenotype
MIM: 607751
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TAS2R38 PubMed Links

Gene Details

Gene
TAS2R38
Gene Name
taste 2 receptor member 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_176817.4 1054 Missense Mutation AAC,GAC N324D NP_789787.4

View Full Product Details