Product Details

SNP ID
rs140442131
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:103354885 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACCTAGGGTCAGAGCACTTACTC[C/T]AGGCAGATCAAGCAAGTTGAAGATG
Phenotype
MIM: 605502 MIM: 154365 MIM: 604943
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJC2 PubMed Links

Gene Details

Gene
DNAJC2
Gene Name
DnaJ heat shock protein family (Hsp40) member C2
There are no transcripts associated with this gene.

Gene
PSMC2
Gene Name
proteasome 26S subunit, ATPase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204453.1 435 Silent Mutation TCC,TCT S42S NP_001191382.1
NM_002803.3 435 Silent Mutation TCC,TCT S42S NP_002794.1
XM_005250505.1 435 Silent Mutation TCC,TCT S42S XP_005250562.1
Gene
SLC26A5
Gene Name
solute carrier family 26 member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167962.1 435 Intron NP_001161434.1
NM_001321787.1 435 Intron NP_001308716.1
NM_198999.2 435 Intron NP_945350.1
NM_206883.2 435 Intron NP_996766.1
NM_206884.2 435 Intron NP_996767.1
NM_206885.2 435 Intron NP_996768.1
XM_011516170.2 435 Intron XP_011514472.1

View Full Product Details