Product Details
- SNP ID
-
rs140513683
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:143139095 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCAGGTTAAAACTTACCTCATTAG[C/T]AGCATCCCTCTACAAGGTGCATTTA
- Phenotype
-
MIM: 176720
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PIP
PubMed Links
Gene Details
- Gene
- PIP
- Gene Name
- prolactin induced protein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002652.2 |
258 |
Silent Mutation |
AGC,AGT |
S74S |
NP_002643.1 |
View Full Product Details