Product Details

SNP ID
rs140768905
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:122995149 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGTTTGTCAGTTACAGTGCTGTT[A/T]CTTGGTAGATGCTCCATGGTGAGTA
Phenotype
MIM: 604867
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TAS2R16 PubMed Links

Gene Details

Gene
TAS2R16
Gene Name
taste 2 receptor member 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016945.2 552 Missense Mutation AGA,AGT R162S NP_058641.1

View Full Product Details