Product Details

SNP ID
hCV172935425
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:75988501 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTAGAGCTTGAACTTCTCATACTC[A/G]TCCTTGTAGGCAAACCTGGGGGGCG
Phenotype
MIM: 124015 MIM: 616695 MIM: 616550
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
POR PubMed Links

Gene Details

Gene
POR
Gene Name
cytochrome p450 oxidoreductase
There are no transcripts associated with this gene.

Gene
STYXL1
Gene Name
serine/threonine/tyrosine interacting-like 1
There are no transcripts associated with this gene.

Gene
TMEM120A
Gene Name
transmembrane protein 120A
There are no transcripts associated with this gene.

View Full Product Details