Product Details

SNP ID
rs140936673
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:41689935 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATGATCCAGTCATTCCAGCCGAT[A/G]TCCTTGAAACTGACAAAGAACTGTT
Phenotype
MIM: 147290
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
INHBA PubMed Links

Gene Details

Gene
INHBA
Gene Name
inhibin beta A subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017012174.1 3003 Silent Mutation GAC,GAT D454D XP_016867663.1
XM_017012175.1 3003 Silent Mutation GAC,GAT D389D XP_016867664.1
XM_017012176.1 3003 Silent Mutation GAC,GAT D332D XP_016867665.1
Gene
INHBA-AS1
Gene Name
INHBA antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details