Product Details

SNP ID
rs141214897
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:149765538 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCTGGCCGGGCCGGCCGTCTGGT[A/G]CACCCTTTGGTGCCGCACCAAGTGC
Phenotype
MIM: 614040
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF467 PubMed Links

Gene Details

Gene
ZNF467
Gene Name
zinc finger protein 467
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005249959.4 1077 Missense Mutation CAC,TAC H322Y XP_005250016.1
XM_005249960.4 1077 Missense Mutation CAC,TAC H322Y XP_005250017.1
XM_005249961.4 1077 Missense Mutation CAC,TAC H322Y XP_005250018.1
XM_006715864.3 1077 Missense Mutation CAC,TAC H363Y XP_006715927.1
XM_011515855.2 1077 Missense Mutation CAC,TAC H573Y XP_011514157.1
XM_011515856.2 1077 Missense Mutation CAC,TAC H573Y XP_011514158.1
XM_011515857.2 1077 Missense Mutation CAC,TAC H536Y XP_011514159.1
XM_011515858.1 1077 Missense Mutation CAC,TAC H322Y XP_011514160.1
XM_017011799.1 1077 Missense Mutation CAC,TAC H497Y XP_016867288.1

View Full Product Details