Product Details
- SNP ID
-
rs141222652
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:155462561 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACGAGTCACAGATCAAGATTTGGTT[C/G]CAGAACAAGCGCGCCAAGATCAAGA
- Phenotype
-
MIM: 131310
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
EN2
PubMed Links
Gene Details
- Gene
- EN2
- Gene Name
- engrailed homeobox 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001427.3 |
1125 |
Missense Mutation |
TTC,TTG |
F292L |
NP_001418.2 |
View Full Product Details