Product Details

SNP ID
rs141258559
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:124747168 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAATCCTCCTTGCTCAGCTGGCGG[A/G]GAACAACTTCTGGAAGTGCTAACAA
Phenotype
MIM: 602583
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR37 PubMed Links

Gene Details

Gene
GPR37
Gene Name
G protein-coupled receptor 37
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005302.3 2248 Missense Mutation CCC,CTC P400L NP_005293.1

View Full Product Details