Product Details

SNP ID
rs141397005
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:6111207 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAATCAGCCTGGCAGCTCCGAGG[C/G]AGTCTCACCTGGAGACATGGATGCA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
USP42 PubMed Links

Gene Details

Gene
USP42
Gene Name
ubiquitin specific peptidase 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032172.2 139 Missense Mutation GCA,GGA A25G NP_115548.1
XM_005249883.4 139 Missense Mutation GCA,GGA A25G XP_005249940.1
XM_006715790.1 139 Missense Mutation GCA,GGA A25G XP_006715853.1
XM_006715791.2 139 Missense Mutation GCA,GGA A25G XP_006715854.1
XM_011515573.1 139 Missense Mutation GCA,GGA A25G XP_011513875.1
XM_011515574.1 139 UTR 5 XP_011513876.1
XM_011515577.1 139 Intron XP_011513879.1
XM_011515578.1 139 UTR 5 XP_011513880.1

View Full Product Details