Product Details

SNP ID
rs141414600
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:27100410 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATATGCATGACAATGCATCCCA[A/G]TGTAATACAAAAATAAAAAGAAAGT
Phenotype
MIM: 142955 MIM: 604685 MIM: 142954
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HOTAIRM1 PubMed Links

Gene Details

Gene
HOTAIRM1
Gene Name
HOXA transcript antisense RNA, myeloid-specific 1
There are no transcripts associated with this gene.

Gene
HOXA1
Gene Name
homeobox A1
There are no transcripts associated with this gene.

Gene
HOXA2
Gene Name
homeobox A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006735.3 1722 UTR 3 NP_006726.1
Gene
HOXA3
Gene Name
homeobox A3
There are no transcripts associated with this gene.

View Full Product Details