Product Details

SNP ID
rs141503569
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100707097 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATCAACATCGCGCTGCAGCTGCA[A/G]GCGGGCAGCTTCGGGTCCTTGCAGG
Phenotype
MIM: 606113
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
POP7 PubMed Links

Gene Details

Gene
POP7
Gene Name
POP7 homolog, ribonuclease P/MRP subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005837.2 529 Silent Mutation CAA,CAG Q89Q NP_005828.2

View Full Product Details