Product Details
- SNP ID
-
rs141594991
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:151033915 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTGCTGGTCCTGGGGGTAGCCGTC[A/G]TGGTGAGGCTTTCCCCACTTCTCCA
- Phenotype
-
MIM: 605464
MIM: 612205
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ABCB8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs4148844] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ABCB8
- Gene Name
- ATP binding cassette subfamily B member 8
- Gene
- ATG9B
- Gene Name
- autophagy related 9B
There are no transcripts associated with this gene.
View Full Product Details